Assay Details
| Gene Symbol: PAFAH1B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258130.1 | NP_001245059.1 | 1536 | 95 | ||
| XM_028835452.1 | XP_028691285.1 | 1614 | 95 | |||
| GenBank mRNA | AY260741.1 | - | 213 | 95 | ||
| JU321627.1 | - | 1331 | 95 | |||
| JU321628.1 | - | 1331 | 95 | |||
| JU471025.1 | - | 1536 | 95 | |||
| JV044538.1 | - | 1267 | 95 | |||
| JV668035.1 | - | 1546 | 95 | |||
More Information
Set Membership: |
|

PAFAH1B1
Genomic Map