Assay Details
| Gene Symbol: COX6B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040281.1 | NP_001035371.1 | 54 | 88 | ||
| GenBank mRNA | AY585854.1 | - | 54 | 88 | ||
| JU338907.1 | - | 177 | 88 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001040281.1 transcripts.
Set Membership: |
|

COX6B1
Genomic Map