Assay Details
| Gene Symbol: UGDH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261103.1 | NP_001248032.1 | 476 | 104 | ||
| XM_015138161.2 | XP_014993647.1 | 268 | 104 | |||
| GenBank mRNA | JU324342.1 | - | 455 | 104 | ||
| JU470762.1 | - | 466 | 104 | |||
| JV045450.1 | - | 476 | 104 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

UGDH
Genomic Map