Assay Details
| Gene Symbol: AP1AR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193936.2 | NP_001180865.1 | 454 | 94 | ||
| XM_028848041.1 | XP_028703874.1 | 484 | 94 | |||
| XM_028848042.1 | XP_028703875.1 | 171 | 94 | |||
| GenBank mRNA | JU321923.1 | - | 482 | 94 | ||
| JU322045.1 | - | 482 | 94 | |||
| JU473745.1 | - | 422 | 94 | |||
| JV044966.1 | - | 417 | 94 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

AP1AR
Genomic Map