Assay Details
| Gene Symbol: VTA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261520.1 | NP_001248449.1 | 431 | 124 | ||
| XM_028846715.1 | XP_028702548.1 | 416 | 124 | |||
| GenBank mRNA | JU334917.1 | - | 425 | 124 | ||
| JU475925.1 | - | 430 | 124 | |||
| JV047321.1 | - | 431 | 124 | |||
More Information
Set Membership: |
|

VTA1
Genomic Map