Assay Details
| Gene Symbol: PSMA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266881.1 | NP_001253810.1 | 490 | 83 | ||
| GenBank mRNA | JU335507.1 | - | 490 | 83 | ||
| JV528251.1 | - | 492 | 83 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PSMA2
Genomic Map