Assay Details
| Gene Symbol: FAM50A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015128555.2 | XP_014984041.1 | 431 | 81 | ||
| XM_015128556.2 | XP_014984042.1 | 431 | 81 | |||
| GenBank mRNA | JU473440.1 | - | 327 | 81 | ||
| JV044817.1 | - | 328 | 81 | |||
| JV700904.1 | - | 361 | 81 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

FAM50A
Genomic Map