Assay Details
| Gene Symbol: RIT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257707.1 | NP_001244636.1 | 244 | 76 | ||
| XM_028837678.1 | XP_028693511.1 | 220 | 76 | |||
| GenBank mRNA | JU320063.1 | - | 244 | 76 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RIT2
Genomic Map