Assay Details
| Gene Symbol: RGS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001082467.4 | XP_001082467.1 | 1031 | 116 | ||
| GenBank mRNA | JU322839.1 | - | 763 | 116 | ||
| JV489709.1 | - | 788 | 116 | |||
| JV529592.1 | - | 865 | 116 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RGS4
Genomic Map