Assay Details
Gene Symbol: TMEM167B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261468.1 | NP_001248397.1 | 246 | 104 | ||
GenBank mRNA | JU320744.1 | - | 246 | 104 | ||
JU473656.1 | - | 248 | 104 | |||
JV045188.1 | - | 246 | 104 | |||
JV566373.1 | - | 238 | 104 | |||
JV638497.1 | - | 250 | 104 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
