Assay Details
| Gene Symbol: MYL12A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257736.1 | NP_001244665.1 | 179 | 69 | ||
| XM_015121628.2 | XP_014977114.1 | 384 | 69 | |||
| GenBank mRNA | JU332489.1 | - | 179 | 69 | ||
| JU332490.1 | - | 179 | 69 | |||
| JU332559.1 | - | 179 | 69 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001257736.1, XM_015121628.2 transcripts.
Set Membership: |
|

MYL12A
Genomic Map