Assay Details
| Gene Symbol: SPTLC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015149486.2 | XP_015004972.1 | 1253 | 76 | ||
| XM_028828181.1 | XP_028684014.1 | 1252 | 76 | |||
| XM_028828182.1 | XP_028684015.1 | 1253 | 76 | |||
| XM_028828183.1 | XP_028684016.1 | 1253 | 76 | |||
| GenBank mRNA | JV044845.1 | - | 1218 | 76 | ||
| JV636257.1 | - | 1213 | 76 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SPTLC3
Genomic Map