Assay Details
| Gene Symbol: PUM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015117806.2 | XP_014973292.2 | 375 | 103 | ||
| XM_015117807.2 | XP_014973293.2 | 416 | 103 | |||
| XM_028835199.1 | XP_028691032.1 | 553 | 103 | |||
| GenBank mRNA | JU478088.1 | - | 355 | 103 | ||
| JV048863.1 | - | 391 | 103 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PUM3
Genomic Map