Assay Details
| Gene Symbol: IGFBP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015151830.2 | XP_015007316.1 | 750 | 65 | ||
| GenBank mRNA | JU334351.1 | - | 539 | 65 | ||
| JV046965.1 | - | 537 | 65 | |||
More Information
Set Membership: |
|

IGFBP6
Genomic Map