Assay Details
| Gene Symbol: TRAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266375.1 | NP_001253304.1 | 816 | 95 | ||
| XM_028852116.1 | XP_028707949.1 | 787 | 95 | |||
| GenBank mRNA | JU333706.1 | - | 721 | 95 | ||
| JV045469.1 | - | 816 | 95 | |||
| JV637315.1 | - | 773 | 95 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TRAM1
Genomic Map