Assay Details
| Gene Symbol: METTL14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261720.1 | NP_001248649.1 | 191 | 131 | ||
| GenBank mRNA | JU335679.1 | - | 278 | 131 | ||
| JU476579.1 | - | 284 | 131 | |||
| JV047778.1 | - | 191 | 131 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

METTL14
Genomic Map