Assay Details
| Gene Symbol: AP3M1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001098843.4 | XP_001098843.2 | 1137 | 135 | ||
| XM_015147273.2 | XP_015002759.1 | 868 | 135 | |||
| XM_015147274.2 | XP_015002760.1 | 826 | 135 | |||
| XM_028826451.1 | XP_028682284.1 | 1251 | 135 | |||
| XM_028826452.1 | XP_028682285.1 | 974 | 135 | |||
| GenBank mRNA | JU320435.1 | - | 886 | 135 | ||
| JU471137.1 | - | 867 | 135 | |||
| JV046754.1 | - | 849 | 135 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

AP3M1
Genomic Map