Assay Details
| Gene Symbol: CCDC126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190944.1 | NP_001177873.1 | 99 | 71 | ||
| GenBank mRNA | CN641731.1 | - | 99 | 71 | ||
| JU321965.1 | - | 227 | 71 | |||
| JV638893.1 | - | 199 | 71 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001190944.1 transcripts.
Set Membership: |
|

CCDC126
Genomic Map