Assay Details
| Gene Symbol: CCDC126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190944.1 | NP_001177873.1 | 565 | 72 | ||
| XM_015133874.2 | XP_014989360.1 | 692 | 72 | |||
| XM_028844986.1 | XP_028700819.1 | 592 | 72 | |||
| GenBank mRNA | CN641731.1 | - | 566 | 72 | ||
| JU321965.1 | - | 693 | 72 | |||
| JU477742.1 | - | 291 | 72 | |||
| JV045723.1 | - | 430 | 72 | |||
| JV638893.1 | - | 665 | 72 | |||
More Information
Set Membership: |
|

CCDC126
Genomic Map