Assay Details
| Gene Symbol: GSK3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266636.2 | NP_001253565.1 | 723 | 66 | ||
| GenBank mRNA | JU327471.1 | - | 559 | 66 | ||
| JV046853.1 | - | 724 | 66 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

GSK3A
Genomic Map