Assay Details
| Gene Symbol: MAP2K3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265947.1 | NP_001252876.1 | 535 | 82 | ||
| GenBank mRNA | JU472916.1 | - | 532 | 82 | ||
| JV043929.1 | - | 535 | 82 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MAP2K3
Genomic Map