Assay Details
| Gene Symbol: ADM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001100827.4 | XP_001100827.1 | 362 | 73 | ||
| XM_015114882.2 | XP_014970368.1 | 362 | 73 | |||
| GenBank mRNA | JV642488.1 | - | 86 | 73 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ADM
Genomic Map