Assay Details
| Gene Symbol: LAMTOR5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001101104.4 | XP_001101104.3 | 1018 | 111 | ||
| GenBank mRNA | JU325047.1 | - | 178 | 111 | ||
| JV640331.1 | - | 247 | 111 | |||
| JV705322.1 | - | 103 | 111 | |||
More Information
Set Membership: |
|

LAMTOR5
Genomic Map