Assay Details
| Gene Symbol: FN3KRP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194815.3 | NP_001181744.2 | 315 | 60 | ||
| GenBank mRNA | JU336402.1 | - | 332 | 60 | ||
| JU477220.1 | - | 300 | 60 | |||
| JV048260.1 | - | 315 | 60 | |||
More Information
Set Membership: |
|

FN3KRP
Genomic Map