Assay Details
Gene Symbol: IFI6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048170.1 | NP_001041635.1 | 71 | 95 | ||
GenBank mRNA | JU323453.1 | - | 67 | 95 | ||
JU332230.1 | - | 67 | 95 | |||
JU332231.1 | - | 67 | 95 | |||
JU473079.1 | - | 89 | 95 | |||
JU473080.1 | - | 89 | 95 | |||
JU473081.1 | - | 89 | 95 | |||
JU473082.1 | - | 89 | 95 | |||
JV046642.1 | - | 94 | 95 | |||
JV046643.1 | - | 94 | 95 | |||
JV046644.1 | - | 94 | 95 | |||
JV046645.1 | - | 94 | 95 | |||
JV046646.1 | - | 94 | 95 | |||
JV046647.1 | - | 94 | 95 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001048170.1 transcripts.
Set Membership: |
![]() ![]() |
