Assay Details
Gene Symbol: IFI6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048170.1 | NP_001041635.1 | 170 | 94 | ||
XM_015132328.2 | XP_014987814.1 | 287 | 94 | |||
XM_028845584.1 | XP_028701417.1 | 117 | 94 | |||
GenBank mRNA | JU323453.1 | - | 166 | 94 | ||
JU332230.1 | - | 166 | 94 | |||
JU332231.1 | - | 166 | 94 | |||
JU473079.1 | - | 188 | 94 | |||
JU473080.1 | - | 188 | 94 | |||
JU473081.1 | - | 188 | 94 | |||
JU473082.1 | - | 188 | 94 | |||
JV046642.1 | - | 193 | 94 | |||
JV046643.1 | - | 193 | 94 | |||
JV046644.1 | - | 193 | 94 | |||
JV046645.1 | - | 193 | 94 | |||
JV046646.1 | - | 193 | 94 | |||
JV046647.1 | - | 193 | 94 |
