Assay Details
| Gene Symbol: CALM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001112409.4 | XP_001112409.3 | 372 | 67 | ||
| XM_015124314.2 | XP_014979800.1 | 469 | 67 | |||
| XM_028839571.1 | XP_028695404.1 | 306 | 67 | |||
| GenBank mRNA | JV044026.1 | - | 238 | 67 | ||
| JV636942.1 | - | 366 | 67 | |||
| JV636943.1 | - | 366 | 67 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CALM3
Genomic Map