Assay Details
| Gene Symbol: YIF1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194498.1 | NP_001181427.1 | 616 | 59 | ||
| XM_015113542.2 | XP_014969028.1 | 654 | 59 | |||
| GenBank mRNA | JU321228.1 | - | 534 | 59 | ||
| JU472347.1 | - | 574 | 59 | |||
| JV043743.1 | - | 491 | 59 | |||
| JV601083.1 | - | 570 | 59 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

YIF1A
Genomic Map