Assay Details
| Gene Symbol: DDIT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257796.1 | NP_001244725.1 | 401 | 68 | ||
| XM_015147370.2 | XP_015002856.1 | 497 | 68 | |||
| GenBank mRNA | JU335091.1 | - | 401 | 68 | ||
| JU476093.1 | - | 429 | 68 | |||
| JV047439.1 | - | 390 | 68 | |||
More Information
Set Membership: |
|

DDIT4
Genomic Map