Assay Details
| Gene Symbol: IFI16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261661.1 | NP_001248590.1 | 1587 | 126 | ||
| GenBank mRNA | JU323500.1 | - | 1587 | 126 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

IFI16| Gene Symbol: IFI16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261661.1 | NP_001248590.1 | 1587 | 126 | ||
| GenBank mRNA | JU323500.1 | - | 1587 | 126 | ||
Set Membership: |
|