Assay Details
| Gene Symbol: IFI16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261661.1 | NP_001248590.1 | 1756 | 70 | ||
| GenBank mRNA | JU323500.1 | - | 1756 | 70 | ||
| JU471830.1 | - | 1786 | 70 | |||
More Information
Set Membership: |
|

IFI16
Genomic Map