Assay Details
| Gene Symbol: METTL2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266641.1 | NP_001253570.1 | 562 | 92 | ||
| GenBank mRNA | JU334314.1 | - | 560 | 92 | ||
| JU334315.1 | - | 560 | 92 | |||
| JU475419.1 | - | 562 | 92 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

METTL2A
Genomic Map