Assay Details
| Gene Symbol: SELENOT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159414.2 | NP_001152886.1 | 701 | 74 | ||
| GenBank mRNA | JU332110.1 | - | 684 | 74 | ||
| JU470706.1 | - | 702 | 74 | |||
| JV703836.1 | - | 689 | 74 | |||
More Information
Set Membership: |
|

SELENOT
Genomic Map