Assay Details
| Gene Symbol: EIF4H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266062.1 | NP_001252991.1 | 262 | 127 | ||
| XM_015133336.2 | XP_014988822.1 | 441 | 127 | |||
| GenBank mRNA | JV600786.1 | - | 262 | 127 | ||
| JV667471.1 | - | 262 | 127 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

EIF4H
Genomic Map