Assay Details
| Gene Symbol: PCNP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001097421.4 | XP_001097421.2 | 474 | 142 | ||
| XM_028844662.1 | XP_028700495.1 | 275 | 142 | |||
| GenBank mRNA | JU322183.1 | - | 379 | 142 | ||
| JU322184.1 | - | 379 | 142 | |||
| JU471536.1 | - | 384 | 142 | |||
| JU471537.1 | - | 384 | 142 | |||
| JV043797.1 | - | 381 | 142 | |||
| JV043798.1 | - | 381 | 142 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PCNP
Genomic Map