Assay Details
| Gene Symbol: ATF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266632.2 | NP_001253561.1 | 921 | 118 | ||
| GenBank mRNA | JU322509.1 | - | 556 | 118 | ||
| JU487167.1 | - | 511 | 118 | |||
| JV043307.1 | - | 630 | 118 | |||
| JV047142.1 | - | 507 | 118 | |||
| JV507789.1 | - | 491 | 118 | |||
| JV636171.1 | - | 921 | 118 | |||
More Information
Set Membership: |
|

ATF4
Genomic Map