Assay Details
| Gene Symbol: CFL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266605.1 | NP_001253534.1 | 508 | 147 | ||
| XM_015113656.2 | XP_014969142.1 | 716 | 147 | |||
| XM_015113657.2 | XP_014969143.1 | 428 | 147 | |||
| XM_015113658.2 | XP_014969144.1 | 1165 | 147 | |||
| XM_028832876.1 | XP_028688709.1 | 612 | 147 | |||
| GenBank mRNA | JV043310.1 | - | 492 | 147 | ||
| JV569273.1 | - | 571 | 147 | |||
| JV635913.1 | - | 508 | 147 | |||
More Information
Set Membership: |
|

CFL1
Genomic Map