Assay Details
| Gene Symbol: SMS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001086702.4 | XP_001086702.3 | 1136 | 121 | ||
| XM_028842153.1 | XP_028697986.1 | 1748 | 121 | |||
| XM_028842154.1 | XP_028697987.1 | 903 | 121 | |||
| XM_028842155.1 | XP_028697988.1 | 864 | 121 | |||
| GenBank mRNA | JU336033.1 | - | 969 | 121 | ||
| JU476898.1 | - | 970 | 121 | |||
| JV048008.1 | - | 977 | 121 | |||
| JV048009.1 | - | 968 | 121 | |||
| JV669578.1 | - | 971 | 121 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SMS
Genomic Map