Assay Details
| Gene Symbol: ARL6IP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257759.1 | NP_001244688.1 | 485 | 144 | ||
| GenBank mRNA | JU332530.1 | - | 485 | 144 | ||
| JU474008.1 | - | 444 | 144 | |||
| JV045936.1 | - | 434 | 144 | |||
| JV705324.1 | - | 508 | 144 | |||
More Information
Set Membership: |
|

ARL6IP5
Genomic Map