Assay Details
| Gene Symbol: NINJ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015150712.2 | XP_015006198.2 | 444 | 119 | ||
| GenBank mRNA | JU344039.1 | - | 170 | 119 | ||
| JV715887.1 | - | 145 | 119 | |||
More Information
Set Membership: |
|

NINJ2
Genomic Map