Assay Details
| Gene Symbol: DCAF16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194742.1 | NP_001181671.1 | 228 | 139 | ||
| XM_015138047.2 | XP_014993533.1 | 882 | 139 | |||
| XM_015138049.2 | XP_014993535.1 | 1196 | 139 | |||
| XM_015138050.2 | XP_014993536.1 | 723 | 139 | |||
| GenBank mRNA | JU321698.1 | - | 410 | 139 | ||
| JU332347.1 | - | 410 | 139 | |||
| JU332348.1 | - | 410 | 139 | |||
| JU332349.1 | - | 410 | 139 | |||
| JU473754.1 | - | 410 | 139 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001194742.1, XM_015138047.2, XM_015138049.2, XM_015138050.2 transcripts.
Set Membership: |
|

DCAF16
Genomic Map