Assay Details
| Gene Symbol: RAB20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001082373.4 | XP_001082373.1 | 712 | 80 | ||
| GenBank mRNA | JV043186.1 | - | 379 | 80 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RAB20| Gene Symbol: RAB20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001082373.4 | XP_001082373.1 | 712 | 80 | ||
| GenBank mRNA | JV043186.1 | - | 379 | 80 | ||
Set Membership: |
|