Assay Details
| Gene Symbol: MSL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266958.1 | NP_001253887.1 | 404 | 62 | ||
| GenBank mRNA | JU320214.1 | - | 731 | 62 | ||
| JU470564.1 | - | 790 | 62 | |||
| JV047513.1 | - | 404 | 62 | |||
| JV638214.1 | - | 881 | 62 | |||
More Information
Set Membership: |
|

MSL2
Genomic Map