Assay Details
| Gene Symbol: GIMAP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257713.1 | NP_001244642.1 | 115 | 136 | ||
| GenBank mRNA | JU335308.1 | - | 115 | 136 | ||
| JU476267.1 | - | 130 | 136 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001257713.1 transcripts.
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

GIMAP4
Genomic Map