Assay Details
| Gene Symbol: GPRC5C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001088824.4 | XP_001088824.2 | 3378 | 79 | ||
| XM_028836761.1 | XP_028692594.1 | 1989 | 79 | |||
More Information
Set Membership: |
|

GPRC5C
Genomic Map