Assay Details
| Gene Symbol: ELAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015121989.2 | XP_014977475.1 | 774 | 101 | ||
| XM_015121990.2 | XP_014977476.1 | 1987 | 101 | |||
| XM_015121991.2 | XP_014977477.1 | 2027 | 101 | |||
| GenBank mRNA | JU322203.1 | - | 690 | 101 | ||
| JU489335.1 | - | 605 | 101 | |||
More Information
Set Membership: |
|

ELAC1
Genomic Map