Assay Details
| Gene Symbol: HOXA10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015133830.2 | XP_014989316.1 | 6243 | 58 | ||
| GenBank mRNA | JU485564.1 | - | 88 | 58 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HOXA10| Gene Symbol: HOXA10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015133830.2 | XP_014989316.1 | 6243 | 58 | ||
| GenBank mRNA | JU485564.1 | - | 88 | 58 | ||
Set Membership: |
|