Assay Details
| Gene Symbol: ACKR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015111240.2 | XP_014966726.1 | 1834 | 169 | ||
| XM_015111241.2 | XP_014966727.1 | 2037 | 169 | |||
| XM_028830379.1 | XP_028686212.1 | 2283 | 169 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ACKR3
Genomic Map