Assay Details
| Gene Symbol: RDH14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001093556.4 | XP_001093556.1 | 495 | 93 | ||
| GenBank mRNA | JU325707.1 | - | 363 | 93 | ||
| JU490552.1 | - | 354 | 93 | |||
More Information
Set Membership: |
|

RDH14
Genomic Map