Assay Details
| Gene Symbol: ZNF500 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001098209.4 | XP_001098209.2 | 203 | 130 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001098209.4 transcripts.
Set Membership: |
|

ZNF500
Genomic Map